Learn More
StatLab BlocFile 1 Corrugated Supa Mega Cassette Storage
Description
- An economical drawer system for routine cassette storage
- Manufactured from high-density fiberboard which ensures its strength, rigidity, and durability
- Made from recycled material
- Drawer system for easy access
- Stackable to save bench space
- Dimensions: 352 x 240 x 60 mm
Specifications
Specifications
| Product Type | Cardboard Block Filing Cabinet |
| Color | Light Brown |
| Holds | 220 Cassettes |