missing translation for 'onlineSavingsMsg'
Learn More

Bleomycin Sulfate (mixture) 85.0+%, TCI America™

Catalog No. B397210MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B397210MG 10 mg
B397250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B397210MG Supplier TCI America Supplier No. B397210MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9041-93-4
Molecular Formula C55H85N17O25S4
Molecular Weight (g/mol) 1512.619
MDL Number MFCD00070310
InChI Key WUIABRMSWOKTOF-UHFFFAOYSA-N
PubChem CID 131664136
IUPAC Name [2-[2-[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[5-[[1-[2-[4-[4-(3-dimethylsulfoniopropylcarbamoyl)-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-3-hydroxy-1-oxobutan-2-yl]amino]-3-hyd
SMILES CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)OC4C(C(C(C(O4)CO)O)OC(=N)[O-])O)C(=O)NC(C)C(C(C)C(=O)NC(C(C)O)C(=O)NCCC5=NC(=CS5)C6=NC(=CS6)C(=O)NCCC[S+](C)C)O.OS(=O)(=O)O

Specifications

Type Bleomycin Sulfate (mixture)
Physical Form Crystalline Powder
Color White/Yellow
Quantity 10 mg
Percent Purity ≥85.0% (HPLC)
Chemical Name or Material Bleomycin Sulfate (mixture)
RTECSNumber EC5991990
TSCA No
Recommended Storage Refrigerator