missing translation for 'onlineSavingsMsg'
Learn More

Blasticidin S Hydrochloride (From Streptomyces Griseochromogenes), Fisher BioReagents

Catalog No. p-24711
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP264725 25 mg Amber Glass
BP264750 50 mg Amber Glass
BP2647100 100 mg Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP264725 Supplier Fisher BioReagents Supplier No. BP264725
Only null left

Chemical Identifiers

CAS 3-9-3513
Molecular Formula C17H27ClN8O5
Molecular Weight (g/mol) 458.90
InChI Key YQXYQOXRCNEATG-UHFFFAOYNA-N
Synonym BLA-S, Cytovirin
PubChem CID 124080960
IUPAC Name (2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
SMILES CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl

Specifications

Type Blasticidin S Hydrochloride
Physical Form Powder/Solid
Melting Point 248°C
Packaging Amber Glass
Potency 900μg/mg min.
Color White
Quantity 25 mg
Assay Percent Range ≥99.0 %
Formula Weight 458.69
Percent Purity 99.0% min.
Chemical Name or Material Blasticidin S Hydrochloride, From Streptomyces griseochromogenes
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May be fatal if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:0
Instability:0
Recommended Storage 0° to 5°C