Learn More
Research Products Corporation Blasticidin S Hydrochloride 10mg/ml Solution, 10 Milligrams
Catalog No. 501368106
Encompass
Blasticidin S HCl Solution 10 mg/mL is a sterile, DNase/RNase-free solution in HEPES buffer. It is a nucleoside antibiotic derived from Streptomyces griseochromogenes with high purity, commonly used in molecular biology, biotechnology, and gene editing applications such as CRISPR. It functions as a selection agent in cell culture by inhibiting protein synthesis in both prokaryotic and eukaryotic cells, allowing for the isolation of successfully modified cells. This product is ideal for creating stable cell lines and expressing recombinant proteins. Store at -20C and avoid repeated freeze-thaw cycles.
Catalog No. 501368106 Supplier Research Products Corporation Supplier No. B121500.01
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Blasticidin S HCl Solution 10 mg/mL is a sterile, DNase/RNase-free solution in HEPES buffer. It is a nucleoside antibiotic derived from Streptomyces griseochromogenes with high purity, commonly used in molecular biology, biotechnology, and gene editing applications such as CRISPR. It functions as a selection agent in cell culture by inhibiting protein synthesis in both prokaryotic and eukaryotic cells, allowing for the isolation of successfully modified cells. This product is ideal for creating stable cell lines and expressing recombinant proteins. Store at -20C and avoid repeated freeze-thaw cycles.