missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blasticidin S HCl, Corning™
Description
- CAS No: 3513-03-9
- Formula: C17H26N8O5 •HCl
- Pure substance/mixture: Substance
Specifications
Specifications
| Chemical Name or Material | Blasticidin S HCl |
| Color | White |
| Form | Powder |
| Product Type | Antibiotic |
| Quantity | 50 mg |