missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Blank Non-Reflective Aluminum Sign

Catalog No. 19129185
Change view
Click to view available options
Color:
White
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19129185 White
19129184 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19129185 Supplier Brady™ Supplier No. 42954

Abrasion-resistant, chemical-resistant, rigid aluminum panel with protected graphics

  • A rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments
  • Non-Reflective Aluminum (B-555) is ideal for indoor and outdoor usage
  • Printed with durable inks and clear-coat protection for fade resistance
  • Withstands wind, rain, sunlight and high temperatures
  • Average outdoor durability 5-8 years
  • Specifications

    Material Aluminum
    Quantity 1 Ea.
    For Use With (Application) Safety Awareness
    Height (English) 10 in.
    Dimensions (L x W) 10 x 14 in.
    Color White