Learn More
Mopec Mounting Bracket for BladeFLASK Blade Remover
Catalog No. AJ151
Change view
Click to view available options
For Use With (Equipment):
Mopec BladeFLASK Blade Remover
Catalog No. For Use With (Equipment)
AJ151 Mopec BladeFLASK Blade Remover
1 options
Catalog No. AJ151 Supplier Mopec Supplier No. AJ151
Mounting bracket for use with Mopec Qlicksmart™ Blade Removal System (AJ150).
  • This bracket provides easy mounting for Mopec BladeFLASK Blade Remover (Cat. No. 22-444-275)
  • Single-handed removal and immediate containment of contaminated scalpel blades dramatically reduces the risk and cost of injury to the user
Specifications
Product Type Mounting Bracket
For Use With (Equipment) Mopec BladeFLASK Blade Remover