Learn More
FEATHER Blade for Replaceable Blade Scissors No.145
Catalog No. 1380425
Change view
Click to view available options
Product Type:
Blunt/Blunt Scissor Blades
Sharp/Blunt Scissor Blades
Sharp/Sharp Scissor Blades
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
1380425 Sharp/Blunt Scissor Blades
1380424 Sharp/Sharp Scissor Blades
1380426 Blunt/Blunt Scissor Blades
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1380425 Supplier FEATHER Supplier No. 1380425
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FEATHER Blades for Replaceable Blade Scissors No.145 come in three different blade types: sharp/sharp, sharp/blunt and blunt/blunt and can all be used with the same handle (sold separately). The replaceable blade design eliminates the cost and trouble of resharpening.
  • Made in Japan
  • Sharp/Sharp or Sharp/Blunt or Blunt/Blunt styles available
  • Stainless steel blades
Specifications
Product Type Sharp/Blunt Scissor Blades
Material Stainless Steel
For Use With (Equipment) Replaceable Blade Scissors No. 145
Quantity Ea.
Width (Metric) 48 mm