Learn More
Mopec Blade Remover, Disposable, 10 pack 
Mopec Blade remover designed to safely remove dull contaminated blade from scalpel handle.
Supplier:  Mopec AJ116
Catalog No. AJ116
Only null left
Explore available promotions
Description
Description
  • Easy to use
  • Autoclavable
  • Holds 100 blades each
  • Not compatible with double sided blades
Specifications
Specifications
Blade Remover
Single-use
10/Pk.
Scalpel Blade
Autoclavable
Product Suggestions
Product Suggestions