Learn More
Epredia™ Blade Holder Packages for the Cryotome™ FSE Cryostat, Low-Profile Cryotome Blade Holder
Hold blades for the Cryotome™ FSE Cryostat with these packages.
Supplier:  Epredia™ SHA78910032
 Afficher plus de versions de ce produit
Numéro de catalogue. 28618356
Il en reste null
Description
Description
Universal Blade Holder:
  • Accepts standard C- or D-profile knife up to 220mm; 120mm is standard length
  • Use with standard steel knives or with disposable blade holder
  • Use in conjunction with disposable blade holders (Part Nos. 40 or 47)
Low-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder for use with low-profile disposable blades
High-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder will accept either standard or heavy-duty high-profile blades, providing a wide range of tissue sectioning options
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS