GA International Blackout Paper Labels 4"" x 1"" / 101.6mm x 25.4mm Rectangle White From -54C to +93C (-65.2F to +199.4F)
Catalog No. 5000571260
Permanent blackout paper labels with a unique opaque design that allows them to obscure any existing printed information, including serial numbers, barcodes, and text. Perfectly suited for general office use, laboratory, and warehouse applications, these cover-up labels are available in roll format and compatible with all standard thermal-transfer barcode printers.
Catalog No.
5000571260
Supplier
GA InternationalSupplier No.
BOP127C32WH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Permanent blackout paper labels with a unique opaque design that allows them to obscure any existing printed information, including serial numbers, barcodes, and text. Perfectly suited for general office use, laboratory, and warehouse applications, these cover-up labels are available in roll format and compatible with all standard thermal-transfer barcode printers.