missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Black Storage Case

Catalog No. 31502292
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
31502292 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31502292 Supplier Synthware™ Supplier No. K130903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Plastic Storage Case for use with Organic Chemistry Kits K161922 and K161420.

  • Convenient case for use with chemistry kits
  • Foam inserts are included

Specifications

Product Type Storage Case
Color Black
Material Plastic
For Use With (Equipment) Organic Chemistry Kit K161922 & K161420