missing translation for 'onlineSavingsMsg'
Learn More

Black Stallion™ Elkskin Leather Welder's Gloves with Nomex™ Lining

Catalog No. 189991314D
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
189991314D X-Large
189991314A Small
189991314B Medium
189991314C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 189991314D Supplier Black Stallion™ Supplier No. 850XL

missing translation for 'validMainframeMsg'

Extra thick, elkskin leather construction provides heat, flame, and abrasion resistance. DuPont™ Nomex™ liner on the back of the glove offers additional fire protection.

  • Elkskin leather construction is extra thick to provide heat, flame, and abrasion resistance
  • DuPont™ Nomex liner offers additional fire-resistant protection
  • Aramid Fiber stitching for spark and abrasion resistance
  • Reversed grain leather palm enhances grip, and provides a softer feel on the hand
  • Thumb strap provides additional reinforcement or high stress areas

Specifications

Color Gold
Material Elkskin Leather
Size X-Large
For Use With (Application) Stick Welding
Glove Cuff Color White
Glove Cuff Style Gauntlet
Liner Material Nomex™
Palm Type Grain Leather
Series 850