Learn More
Black Stallion™ 45 oz. PBI Jumbo Cover Mitts
Catalog No. 19056609
Change view
Click to view available options
Universal
Catalog No.
19056609 Universal
1 options
Catalog No. 19056609 Supplier Black Stallion™ Supplier No. HPC3
Mitts are constructed of 45 oz. PBI for protection during high temperature applications.
  • Heavyweight PBI construction provides thermal insulation during high temperature applications
  • Designed to provide additional protection when worn over another pair of mitts
Specifications
Color Orange
Material PBI
Size Universal
Glove Cuff Style Open
Yarn Weight 45 oz.