Learn More
Black Stallion™ Aluminized Heat-Resistant Coats and Pants
Catalog No. p-26898
Change view
Click to view available options
Size:
38
42
44
46
50
52
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19056877 50
19056851 42
19056509 46
19056635 38
19057786 52
19057787 44
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19056877 Supplier Black Stallion™ Supplier No. 130AHSXLG
Only null left
  • Together, these aluminized garments protect trunk, arms and legs from radiant heat and molten splash
  • Coat, 30 in. long (76cm), features mandarin collar and snap-front closure
  • Also has inside breast pocket and set-in sleeves with open cuffs
  • Pants have reinforced waistband and flame-retardant hook-and-loop fly
  • 19 oz. aluinized carbon/Kevlar
Specifications
Size 50
Material Carbon Dupont™ Kevlar™ aramid fibers
Color Silver