Learn More
USA Scientific Black light absorbing film, non-sterile
Catalog No. NC9009251
Encompass
Black vinyl light absorbing film, 200 um thick, -40C to 80C, non-sterile, 50/box
Catalog No. NC9009251 Supplier USA Scientific Supplier No. 29250500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Black vinyl light absorbing film, 200 um thick, -40C to 80C, non-sterile, 50/box