missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Black Phenolic PolyCone Lined Caps

Catalog No. 02991076
Change view
Click to view available options
For Use With (Equipment):
Wide mouth bottle
Neck Finish:
22-400
24-400
28-400
33-400
No. per Case:
2200 Cs.
3100 Cs.
5000 Cs.
576 Cs.
Packaging:
Bagged
Bulk
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Neck Finish No. per Case Packaging
02991076 Wide mouth bottle 22-400 5000 Cs. Bulk
361027005 Wide mouth bottle 24-400 576 Cs. Bagged
361012194 Wide mouth bottle 33-400 576 Cs. Bagged
02991174 Wide mouth bottle 28-400 3100 Cs. Bulk
02991097 Wide mouth bottle 33-400 2200 Cs. Bulk
028835J Wide mouth bottle 28-400 576 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 02991076 Supplier Qorpak™ Supplier No. CAP00154
Only null left

Forms an exceptionally tight seal and offer a good chemical barrier. Qorpak™ Black Phenolic PolyCone Lined Caps is used for everything from liquids, to soaps, to macro-invertebrates.

  • Molded liner around the sealing areas eliminates leakage, evaporation, contamination, binding and backing off
  • Available in ribbed style
  • Available in bulk/bagged packaging

Specifications

Color Black
Material Phenolic Resin
No. per Case 5000 Cs.
Packaging Bulk
Sterility Non-sterile
Liner Material Polycone
Neck Finish 22-400
Disposable Reusable
Product Type Lined Cap (Ribbed)
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less