Learn More
Qorpak™ Black Phenolic PolyCone Lined Caps
Forms an exceptionally tight seal and offer a good chemical barrier. Qorpak™ Black Phenolic PolyCone Lined Caps is used for everything from liquids, to soaps, to macro-invertebrates.
Supplier: Qorpak™ CAP00195
Description
- Molded liner around the sealing areas eliminates leakage, evaporation, contamination, binding and backing off
- Available in ribbed style
- Available in bulk/bagged packaging
Specifications
| Black | |
| 576 Cs. | |
| Bagged | |
| 28-400 | |
| Wide mouth bottle |
| Phenolic Resin | |
| 12/Pk. | |
| Polycone | |
| Lined Cap (Ribbed) |