Learn More
Qorpak™ Black Phenolic F217 and PTFE Lined Caps
Ideal for use with a wide range of chemicals including analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals and diagnostic reagents. Qorpak™ Black Phenolic F217 and PTFE Lined Caps offer good chemical and temperature resistance.
$1,208.97 - $5,039.44
Specifications
Color Black
Material Phenolic Resin
Sterility Non-sterile
Liner Material F217, PTFE
Disposable Reusable
View More Specs
Products 4
Catalog Number Mfr. No. Neck Finish Product Type No. per Case Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Neck Finish Product Type No. per Case Packaging Price Quantity & Availability  
02991112
View Documents Promotion Details
Qorpak™
CAP00191
28-400 Lined Cap (Ribbed) 2800 Bulk
Case of 3200 Each for $5,039.44
Only null left
02993373
View Documents Promotion Details
Qorpak™
CAP06691
24-400 Lined Cap (Ribbed) 576 Bagged
Case of 576 Each for $1,208.97
Only null left
02991058
View Documents Promotion Details
Qorpak™
CAP00240
38-430 Lined Cap (Ribbed) 840 Bulk
Case of 840 Each for $2,160.66
Only null left
02991020
View Documents Promotion Details
Qorpak™
CAP07459
38-439 Lined Cap (Ribbed) 840 Bulk
Case of 804 Each for $2,490.13
Only null left
Description
Description
  • Adherence consistency to close dimensional tolerances
  • Resist from chemical attacks at room temperature
  • Provide resilient tight seal with low moisture transmission rate
  • Available in ribbed or smooth style
  • Available in bagged or bulk packaging
Specifications
Specifications
Black
Non-sterile
Reusable
Phenolic Resin
F217, PTFE
Not Autoclavable