Learn More
Fisherbrand™ Black Phenolic Caps
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
 View more versions of this product
Catalog No. FB02911803
Only null left
Description
Description
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Chemical Identifiers
Black
Polyethylene Cone
Phenolic
Specifications
Black
Black
No
Phenolic
5500
20-400
Polyethylene Cone
Product Suggestions
Product Suggestions