Learn More
Fisherbrand™ Black Phenolic Caps
Catalog No. FB02911727A
Change view
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
1600
3100
5500
576
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Quantity
FB02911727A 38-400 1600
FB02912054 20-400 576
FB02911803 20-400 5500
FB02912055 24-400 12
FB02912056 22-400 12
FB02912059 18-400 12
FB02912058 15-425 576
FB02912061 28-400 576
FB02912062 33-400 12
FB02912101 28-400 3100
FB02912063A 38-400 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. FB02911727A
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Color Black
Quantity 1600
Closure Color Black
Neck Finish 38-400
Closure Included No
Closure Liner Polyethylene Cone
Closure Material Phenolic