Learn More
Alkali Scientific ASI™ Black Confocal Dish 35 x10mm, Easy-Grip, Adhesion Type, Sterile, Non-Treated (Non-Charged/ Hydrophobic), 100/Cs, 22 x 22 x 0.13mm, Hole 1.33mm, PS/Glass
Catalog No. 503028467
Encompass
ASI™ confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products:Glass:naturally charged, good growth surface for adherent cellsFLux:surface modified plastic film (188 m thick), optimal for confocal microscopy Bottom materials: Glass/FLux (no surface treatment for glass bottom) Coverslip: #1.5 Dish color: Clear/Black Dish size: 35 Hole size: 13 / 20 / 25 Non pyrogenic Non cytotoxic DNase / RNase free Human DNA free
Catalog No. 503028467 Supplier Alkali Scientific Supplier No. TCG24
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ASI™ confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products:Glass:naturally charged, good growth surface for adherent cellsFLux:surface modified plastic film (188 m thick), optimal for confocal microscopy Bottom materials: Glass/FLux (no surface treatment for glass bottom) Coverslip: #1.5 Dish color: Clear/Black Dish size: 35 Hole size: 13 / 20 / 25 Non pyrogenic Non cytotoxic DNase / RNase free Human DNA free