missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Conductive Automation Tips for Qiagen™ Workstations

Catalog No. p-8636011
Change view
Click to view available options
Volume (Metric):
300 μL
1000 μL
1100 μL
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Sterile
Sterile and PCR Ready
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Fit for Purpose
21402353 1000 μL Filtered Sterile and PCR Ready
21402354 1100 μL Non-filtered Sterile
21402352 300 μL Non-filtered Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21402353 Supplier Thermo Scientific™ Supplier No. 953011
Only null left

Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.

  • Available in nonsterile, sterile and with the ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Black
Format Rack
Tip Style Conductive
Packaging 96 Tips/Tray, 10 Trays/Case
For Use With (Equipment) Universal Fit, Qiagen
Product Line Qiagen
Fit for Purpose Sterile and PCR Ready