missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Black 96-Well Immuno Plates
GREENER_CHOICE

Catalog No. 21377271
Change view
Click to view available options
Surface Treatment:
High Binding
MaxiSorp
Medium Binding
PolySorp
Universal Binding
Volume (Metric) Working:
280 μL
350 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Volume (Metric) Working
21377271 Universal Binding 350 μL
1256624 MaxiSorp 350 μL
1256630 PolySorp 350 μL
14245197A Medium Binding 280 μL
14245177 High Binding 280 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21377271 Supplier Thermo Scientific™ Supplier No. 9502867

Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with these black, 96-well immuno plates, available in a choice of three surface types.

Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.

Features:

  • Plate has SBS footprint — fits standard equipment
  • Flat-bottom wells
  • Certified binding homogeneity/reproducibility

Choice of surface:

  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
  • Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.

Compatible with:

Compatible with lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane

Ordering Information:

Availability may vary by country

Specifications

Binding Property Hydrophobic
Format Standard
Lid Without Lid
Detection Method Fluorescence
Array 8 x 12
Packaging 50 Cases
Binding Type Adsorption
Color Black
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Volume (Metric) Working 350 μL
Sterility Non-sterile
Surface Treatment Universal Binding
Well Shape Flat Bottom (F-well)
For Use With (Application) Fluorescence
No. per Case 50
No. per Pack 25
Plate Blocking None
Quantity 50/Cs.
Target Molecule Lipids
Show More Show Less