missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BL21(DE3) Competent Cells

Catalog No. ferec0114
Change view
Click to view available options
Quantity:
20 Reactions (10 x 100 μL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREC0114 20 Reactions (10 x 100 μL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREC0114 Supplier Thermo Scientific™ Supplier No. EC0114
Only null left

Suitable for the expression of non-toxic heterologous genes. The strain contains the lambda DE3 prophage that carries the gene for T7 RNA polymerase under control of a lacUV5 promoter, allowing expression of the T7 RNA polymerase to be induced with IPTG.

  • Suitable for the expression of non-toxic heterologous genes
  • The strain contains the lambda DE3 prophage that carries the gene for T7 RNA polymerase under control of a lacUV5 promoter, allowing expression of the T7 RNA polymerase to be induced with IPTG
  • BL21(DE3) is an E. coli B strain and does not contain the lon protease. It is also deficient in the outer membrane protease OmpT. The lack of these two key proteases reduces degradation of heterologous proteins expressed in the cells.
  • High transformation efficiency: >1x107 cfu/μg pUC19 DNA
  • Specifically constructed for high-level expression of recombinant proteins
  • Genetic markers that minimize RNA and protein degradation
  • Convenient two reactions per vial packaging
  • S.O.C. Outgrowth medium included
  • Applications:  BL21(DE3) Competent Cells are suitable for high-level T7 expression of recombinant proteins
  • Genotype: F– ompT hsdSB (rB–, mB–) gal dcm (DE3)

Note: Not suitable for maintenance of expression plasmids. Use DH10B or DH5α competent cells that do not carry the gene for T7 RNA polymerase for plasmid DNA propagation (Cat. Nos. EC0112, EC0113).

Specifications

Description Chemically Competent
Contains F' Episome Lacks F' Episome
Content And Storage Dry Ice (-80°C)
Bacterial or Yeast Strain BL21(DE3)
Species E. coli
Format Tube
For Use With (Application) Cloning
Quantity 20 Reactions (10 x 100 μL)