Learn More
Addexbio Technologies BJAB Cells, Frozen Vial
Catalog No. NC1615348
Encompass
Established from the inguinal tumor of an EBV-negative 5-year-old African girl with Burkitt lymphoma at the preterminal stage.
Catalog No. NC1615348 Supplier Addexbio Technologies Supplier No. C0003016
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Established from the inguinal tumor of an EBV-negative 5-year-old African girl with Burkitt lymphoma at the preterminal stage.