missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Biuret, 97%, extra pure

Catalog No. AC106520050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106520050 5 g Glass bottle
AC106520250 25 g Glass bottle
AC106521000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106520050 Supplier Thermo Scientific Chemicals Supplier No. 106520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-19-0 | C2H5N3O2 | 103.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.08
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea
SMILES C(=O)(N)NC(=O)N

Specifications

Melting Point 187°C to 194°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex N)
Packaging Glass bottle
Linear Formula H2NCONHCONH2
Quantity 5 g
Beilstein 03,7
Merck Index 15,131
Solubility Information Solubility in water: 2g/100mL water (25°C). Other solubilities: freely soluble in alcohols,slightly soluble in diethyl ether
Formula Weight 103.08
Percent Purity 97%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Biuret
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-559-

RUO – Research Use Only