missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biurea, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 118.10 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Biurea |
Safety and Handling
| EINECSNumber | (2)-1748 |
| TSCA | Yes |