missing translation for 'onlineSavingsMsg'
Learn More

Bispyrazolone, For TLC derivatization, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111015426
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015426 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015426 Supplier Merck Emd Millipore Supplier No. 151565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00020765
Synonym 3,3 ′-Dimethyl-1,1 ′-diphenyl-(4,4 ′-bi-2-pyrazoline)-5,5 ′-dione

Specifications

CAS 7477-67-0
Linear Formula C20H18N4O2
Quantity 5 g
Solubility Information Formic acid: 50 mg/mL, clear, yellow
Formula Weight 346.38
Percent Purity ≥98.0% (TLC); ≥98.0%
Grade For TLC derivatization