missing translation for 'onlineSavingsMsg'
Learn More

Bispyrazolone, 98+%

Catalog No. p-7021816
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAA1430806 5 g
AAA1430814 25 g
2 options

Catalog No. AAA1430806

Supplier: Thermo Scientific Chemicals A1430806

Only null left

CAS: 7477-67-0 | C20H18N4O2 | 346.39 g/mol

It is applied as a reagent for the detection and determination of cyanide. It is also used in an experimental procedure, done in order to remove linamarin in starch from cassava roots, by using plant cell wall-degrading enzymes, xylanase and cellulase. It was also used for spectrophotometric determination and quantization of urea in urine samples. It is also used in assay of cyanogenic compounds and determination of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reagent for the detection and determination of cyanide. It is also used in an experimental procedure, done in order to remove linamarin in starch from cassava roots, by using plant cell wall-degrading enzymes, xylanase and cellulase. It was also used for spectrophotometric determination and quantization of urea in urine samples. It is also used in assay of cyanogenic compounds and determination of ammonia.

Solubility
Solubility in hot DMF is almost transparent. Soluble in formic acid.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 7477-67-0
Molecular Formula C20H18N4O2
Molecular Weight (g/mol) 346.39
MDL Number MFCD00020765
InChI Key FORCWSNQDMPPOC-UHFFFAOYSA-N
Synonym bispyrazolone, 4,4'-bis 1-phenyl-3-methyl-5-pyrazolone, 4,4'-bi-3h-pyrazole-3,3'-dione, 2,2',4,4'-tetrahydro-5,5'-dimethyl-2,2'-diphenyl, 4,4'-bis 3-methyl-1-phenyl-5-pyrazolone, 4,4'-bi-2-pyrazoline-5,5'-dione, 3,3'-dimethyl-1,1'-diphenyl, 3,3'-dimethyl-1,1'-diphenyl-1h,1'h-4,4'-bipyrazole-5,5' 4h,4'h-dione, 5-methyl-4-3-methyl-5-oxo-1-phenyl-4h-pyrazol-4-yl-2-phenyl-4h-pyrazol-3-one, 4,5'-dione, 3,3'-dimethyl-1,1'-diphenyl
PubChem CID 95807
IUPAC Name 5-methyl-4-(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)-2-phenyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1C2C(=NN(C2=O)C3=CC=CC=C3)C)C4=CC=CC=C4

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 572787
Formula Weight 346.39
Percent Purity ≥98%
Chemical Name or Material Bispyrazolone
Hazard Category Warning
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-282-5
TSCA Yes

RUO – Research Use Only