missing translation for 'onlineSavingsMsg'
Learn More

Bismuthiol 98.0+%, TCI America™

Catalog No. D062225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D062225G 25 g
D0622500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D062225G Supplier TCI America Supplier No. D062225G

Chemical Identifiers

CAS 1072-71-5
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
MDL Number MFCD00003103
InChI Key BIGYLAKFCGVRAN-UHFFFAOYSA-N
Synonym 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i
PubChem CID 2723630
IUPAC Name 1,3,4-thiadiazolidine-2,5-dithione
SMILES S=C1NNC(=S)S1

Specifications

Melting Point 156°C
Quantity 25 g
Formula Weight 150.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bismuthiol
EINECSNumber (5)-0566
RTECSNumber XI3850000
TSCA Yes