missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Bismuthiol | |
| C2H2N2S3 | |
| 500 g | |
| BIGYLAKFCGVRAN-UHFFFAOYSA-N | |
| 1,3,4-thiadiazolidine-2,5-dithione | |
| 2723630 | |
| ≥95.0% (T) |
| 1072-71-5 | |
| MFCD00003103 | |
| 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i | |
| S=C1NNC(=S)S1 | |
| 150.23 | |
| 150.23 | |
| Crystalline Powder |
Chemical Identifiers
| 1072-71-5 | |
| 150.23 | |
| BIGYLAKFCGVRAN-UHFFFAOYSA-N | |
| 2723630 | |
| S=C1NNC(=S)S1 |
| C2H2N2S3 | |
| MFCD00003103 | |
| 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i | |
| 1,3,4-thiadiazolidine-2,5-dithione |
Safety and Handling
EINECSNumber : (5)-0566
RTECSNumber : XI3850000
TSCA : Yes