missing translation for 'onlineSavingsMsg'
Learn More

Bismuth tungsten oxide, 99.9% (metals basis)

Catalog No. AA4185422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4185422 100 g
AA4185414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4185422 Supplier Thermo Scientific Chemicals Supplier No. 04185422
Only null left

CAS: 13595-87-4 | Bi2O12W3 | 1161.469 g/mol

Acts as a visible-light driven photocatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13595-87-4
Molecular Formula Bi2O12W3
Molecular Weight (g/mol) 1161.469
MDL Number MFCD00046190
InChI Key DWNBMQNJXHTBPG-UHFFFAOYSA-N
Synonym bismuth tungstate, bismuth tungsten oxide, 2bi.3wo4, dibismuth 3+ ion tritungstate, dibismuth;dioxido dioxo tungsten, dibismuth 3+ tritungstate, bismuth tungsten oxide, powder,-200 mesh trace metals basis
PubChem CID 22234745
IUPAC Name dibismuth;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[O-][W](=O)(=O)[O-].[O-][W](=O)(=O)[O-].[Bi+3].[Bi+3]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 1161.48
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Bismuth tungsten oxide
EINECSNumber 237-033-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only