missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth tungsten oxide, 99.9% (metals basis)
Description
Acts as a visible-light driven photocatalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 25 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 1161.48 |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Assay | (metals basis) |
| Chemical Name or Material | Bismuth tungsten oxide |
Safety and Handling
| EINECSNumber | 237-033-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only