missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subsalicylate, Purified, 56.5-60.5%, Spectrum™ Chemical

Catalog No. 18609118
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609118 125 g Amber Glass Bottle
18609119 500 g Amber Glass Bottle
18609120 2.5 kg Amber Glass Bottle
18609121 12 kg Amber Glass Bottle
18609122 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609118 Supplier Spectrum Chemical Mfg Cor Supplier No. BI152125GM

Chemical Identifiers

CAS 14882-18-9

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 125 g
Percent Purity 56.5 to 60.5%
Grade Purified
Loss on Drying 0.01
Packaging Amber Glass Bottle
Chemical Name or Material Bismuth Subsalicylate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.