missing translation for 'onlineSavingsMsg'
Learn More

Bismuth subsalicylate, 96%

Catalog No. p-4407273
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4039530 250 g
AA40395A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4039530 Supplier Thermo Scientific Chemicals Supplier No. 04039530

CAS: 14882-18-9 | C7H6BiO4 | 363.101 g/mol

To impart pearly surface to cellulose-base, polystyrene, and phenolformaldehyde resins. Typical tap bulk density 0.43g/cm^3Bismuth subsalicylate is used to treat temporary discomforts of the stomach and gastrointestinal tract, such as diarrhea, indigestion, heartburn and nausea. It also acts as an antacid. It is practiced to decrease the number of bowel movements and to make the stool firmer. It is also used to reduce inflammation within the intestine. It is further used in growth media for selective isolation of microorganisms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14882-18-9
Molecular Formula C7H6BiO4
Molecular Weight (g/mol) 363.101
MDL Number MFCD00085368
InChI Key QBWLKDFBINPHFT-UHFFFAOYSA-L
Synonym bismuth subsalicylate, bismuth oxysalicylate, wismutsubsalicylat, bismuth oxide salicylate, stabisol, vismut, bismuthi subsalicylas, spiromak forte, pepto-bismol, bismutum subsalicylicum
PubChem CID 16682734
IUPAC Name 1,3,2$l^{2}-benzodioxabismin-4-one;hydrate
SMILES C1=CC=C2C(=C1)C(=O)O[Bi]O2.O

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 96%
Solubility Information Practically insoluble in water,alcoholSoluble in acid and alkali. Insoluble in cold water,ethanol,diethyl ether and n-octanol.
Formula Weight 362.09
Odor Odorless
Appearance White powder
Chemical Name or Material Bismuth subsalicylate
Hazard Category H315-H319-H335
EINECSNumber 238-953-1
RTECSNumber EB2985000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only