missing translation for 'onlineSavingsMsg'
Learn More

Bismuth subgallate hydrate

Catalog No. p-7027438
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2418722 100 g
AAB2418736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2418722 Supplier Thermo Scientific Chemicals Supplier No. B2418722
Only null left

CAS: 342406-26-2 | C7H5BiO6 | 394.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 342406-26-2
Molecular Formula C7H5BiO6
Molecular Weight (g/mol) 394.09
MDL Number MFCD00044980
InChI Key JAONZGLTYYUPCT-UHFFFAOYSA-K
Synonym bismuth subgallate, wismutgallathydroxid, dermatol, bismuth gallate, dermatol puder, bismuth subgallas, bismutum subgallicum, basic bismuth gallate, gallic acid bismuth basic salt, wismutgallat, basisches
PubChem CID 16682999
IUPAC Name 4-hydroxy-1,3,2$l^{2}-benzodioxabismole-6-carboxylic acid;hydrate
SMILES O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O

Specifications

Quantity 100 g
Formula Weight 394.10 (Anhydrous)
Chemical Name or Material Bismuth subgallate hydrate
EINECSNumber 202-742-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only