missing translation for 'onlineSavingsMsg'
Learn More

Bismuth rod, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

$1083.90 - $1083.90

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA47229KF
View Documents
Thermo Scientific Chemicals
047229KF
1 Pc.
Each for $1,083.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
CHEBI:33301
[Bi]

Specifications

7440-69-9
Odorless
Bi
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
[Bi]
208.98
CHEBI:33301
Puratronic
(metals basis)
1 Pc.
≈7.5g/cm
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
5359367
99.9999%
Bismuth rod, 10mm (0.39 in.) dia. x 145mm (5.71 in.) long
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-177-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only