missing translation for 'onlineSavingsMsg'
Learn More

Bismuth pieces, 10cm (3.9in) & down, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. AA1220830
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1220830 250 g
AA12208A1 1 kg
AA12208A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1220830 Supplier Thermo Scientific Chemicals Supplier No. 01220830

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Color Gray
Appearance Gray needles
Quantity 250 g
Odor Odorless
Vapor Pressure 1 hPa (1mm Hg)
Form Pieces
Assay Percent Range 99.99% (Metals basis)
Chemical Name or Material Bismuth
EINECSNumber 231-177-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only