missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth needles, 99.99% (metals basis), Thermo Scientific Chemicals
Form: Approx. 1.5-3.0mm diameter; Various lengths
Supplier: Thermo Scientific Chemicals 04148536
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gray | |
| 7440-69-9 | |
| 9.79g/cm3 | |
| Odorless | |
| Needle | |
| MFCD00134033 | |
| JCXGWMGPZLAOME-UHFFFAOYSA-N | |
| bismuth | |
| 5359367 |
| Bismuth | |
| Gray needles | |
| 500 g | |
| 99.99% (Metals basis) | |
| Bi | |
| bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
| [Bi] | |
| 208.98 | |
| CHEBI:33301 |
Chemical Identifiers
| 7440-69-9 | |
| 208.98 | |
| JCXGWMGPZLAOME-UHFFFAOYSA-N | |
| 5359367 | |
| bismuth |
| Bi | |
| MFCD00134033 | |
| bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
| CHEBI:33301 | |
| [Bi] |
Safety and Handling
EINECSNumber : 231-177-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only