missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) trifluoromethanesulfonate, 99%

CAS: 88189-03-1 | C3BiF9O9S3 | 656.17 g/mol

$133.81 - $466.04

Chemical Identifiers

CAS 88189-03-1
Molecular Formula C3BiF9O9S3
Molecular Weight (g/mol) 656.17
MDL Number MFCD02093669
InChI Key NYENCOMLZDQKNH-UHFFFAOYSA-K
Synonym bismuth iii trifluoromethanesulfonate, bismuth triflate, bismuth trifluoromethanesulfonate, bismuth 3+ triflate, bismuth triflate mi, bismuth 3+ trifluoromethanesulfonate, bismuth tris trifluoromethanesulfonate, bismuth iii triflate, bismuth 3+ ion tritriflate
PubChem CID 9917655
IUPAC Name bismuth;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Bi+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1968706
View Documents
Thermo Scientific Chemicals
L1968706
5 g N/A
Only null left
 
AAL1968714
View Documents
Thermo Scientific Chemicals
L1968714
25 g N/A
Only null left
 
Description

Description

Bismuth(III) trifluoromethanesulfonate acts as a catalyst in Friedel-Crafts acylation and cycloisomerization of allene-enol ethers. It behaves as a direct substitution catalyst and involved in the substitution of allylic, propargylic, and benzylic alcohols with sulfonamides, carboxamides and carbamates. Further, it is also used in Mukaiyama aldol reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) trifluoromethanesulfonate acts as a catalyst in Friedel-Crafts acylation and cycloisomerization of allene-enol ethers. It behaves as a direct substitution catalyst and involved in the substitution of allylic, propargylic, and benzylic alcohols with sulfonamides, carboxamides and carbamates. Further, it is also used in Mukaiyama aldol reactions.

Solubility
Soluble in organics acetonitrile, dioxane, dimethyl formamide and dimethyl sulfoxide.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

88189-03-1
656.17
NYENCOMLZDQKNH-UHFFFAOYSA-K
9917655
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Bi+3]
C3BiF9O9S3
MFCD02093669
bismuth iii trifluoromethanesulfonate, bismuth triflate, bismuth trifluoromethanesulfonate, bismuth 3+ triflate, bismuth triflate mi, bismuth 3+ trifluoromethanesulfonate, bismuth tris trifluoromethanesulfonate, bismuth iii triflate, bismuth 3+ ion tritriflate
bismuth;trifluoromethanesulfonate

Specifications

88189-03-1
C3BiF9O9S3
MFCD02093669
UN3261
bismuth iii trifluoromethanesulfonate, bismuth triflate, bismuth trifluoromethanesulfonate, bismuth 3+ triflate, bismuth triflate mi, bismuth 3+ trifluoromethanesulfonate, bismuth tris trifluoromethanesulfonate, bismuth iii triflate, bismuth 3+ ion tritriflate
NYENCOMLZDQKNH-UHFFFAOYSA-K
bismuth;trifluoromethanesulfonate
9917655
99%
>300°C
(CF3SO3)3Bi
5 g
Hygroscopic
Soluble in organics acetonitrile,dioxane,dimethyl formamide and dimethyl sulfoxide.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Bi+3]
656.17
656.19
Bismuth(III) trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only