missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxychloride, 97%, pure

Catalog No. p-7077440
Change view
Click to view available options
Quantity:
50 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453840500 50 g
AC453840010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453840500 Supplier Thermo Scientific Chemicals Supplier No. 453840500
Only null left

CAS: 7787-59-9 | BiClO | 260.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-59-9
Molecular Formula BiClO
Molecular Weight (g/mol) 260.43
InChI Key BWOROQSFKKODDR-UHFFFAOYSA-N
Synonym bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid
PubChem CID 6328152
IUPAC Name oxobismuth;hydrochloride
SMILES O=[Bi].Cl

Specifications

Quantity 50 g
Assay Percent Range 77.0 to 82.5% (Bi)
Linear Formula O=BiCl
Merck Index 15,1274
Solubility Information Solubility in water: insoluble.
Formula Weight 260.43
Percent Purity 97%
Grade Pure
Chemical Name or Material Bismuth(III) oxychloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 232-122-7

RUO – Research Use Only