missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048478
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
AA1065714 25 g
AA1065722 100 g
2 options

Catalog No. AA1065714

Supplier: Thermo Scientific Chemicals 01065714

Only null left

CAS: 10361-44-1 | BiN3O9 | 394.99 g/mol

It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10361-44-1
Molecular Formula BiN3O9
Molecular Weight (g/mol) 394.99
MDL Number MFCD00003460
InChI Key PPNKDDZCLDMRHS-UHFFFAOYSA-N
Synonym bismuth nitrate, bismuth trinitrate, ccris 7110, nitric acid, bismuth 3+ salt, tris nitrooxy bismuthine, bismuth 3+ trinitrate, bi.3no3, bismuth iii nitrate, bismuth iii nitrate hydrate, nitric acid, bismuth 3+ salt 3:1
PubChem CID 107711
IUPAC Name bismuth;trinitrate
SMILES [Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 30°C (decomposition)
Color Colorless to White
Physical Form Crystalline
Quantity 25 g
UN Number UN1477
Merck Index 14,1271
Solubility Information Slightly soluble in water; soluble in acetone,acetic acid,and glycerol. Insoluble in ethyl acetate and ethanol.
Formula Weight 394.99 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Bismuth(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 233-791-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only