missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) fluoride, anhydrous, 99%

Manufacturing of bismuth pentafluoride | CAS: 7787-61-3 | BiF3 | 265.976 g/mol

$98.49 - $926.78

Chemical Identifiers

CAS 7787-61-3
Molecular Formula BiF3
Molecular Weight (g/mol) 265.976
MDL Number MFCD00010893
InChI Key BRCWHGIUHLWZBK-UHFFFAOYSA-K
Synonym bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
PubChem CID 82233
IUPAC Name trifluorobismuthane
SMILES F[Bi](F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1307514
View Documents
Thermo Scientific Chemicals
01307514
25 g
Each for $98.49
Only null left
 
AA1307522
View Documents
Thermo Scientific Chemicals
01307522
100 g
Each for $255.97
Only null left
 
AA1307536
View Documents
Thermo Scientific Chemicals
01307536
500 g
Each for $926.78
Only null left
 
Description

Description

It is used in the manufacturing of bismuth pentafluoride. Bismuth(III) fluoride is an attractive reagent and catalyst in organic synthesis owing to its low cost, and ease of handling. It is also used in polarography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7787-61-3
265.976
BRCWHGIUHLWZBK-UHFFFAOYSA-K
82233
F[Bi](F)F
BiF3
MFCD00010893
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
trifluorobismuthane

Specifications

727°C
Powder
BiF3
UN3260
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
BRCWHGIUHLWZBK-UHFFFAOYSA-K
trifluorobismuthane
82233
99%
1.74
Bismuth(III) fluoride, Anhydrous
7787-61-3
25 g
MFCD00010893
14,1265
Insoluble in water; Soluble in HF.
F[Bi](F)F
265.976
265.98
Odorless
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 232-124-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only