Learn More
Bismuth(III) chloride oxide, 98%
Used as pigment, in manufacture of artificial pearls, dry-cell cathodes | CAS: 7787-59-9 | BiClHO | 261.437 g/mol
$108.04 - $1237.48
Chemical Identifiers
| CAS | 7787-59-9 |
|---|---|
| Molecular Formula | BiClHO |
| Molecular Weight (g/mol) | 261.437 |
| MDL Number | MFCD00010895 |
| InChI Key | BWOROQSFKKODDR-UHFFFAOYSA-N |
| Synonym | bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid |
| PubChem CID | 6328152 |
| IUPAC Name | oxobismuth;hydrochloride |
| SMILES | O=[Bi].Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4039618
|
Thermo Scientific Chemicals
04039618 |
50 g |
Each for $108.04
|
|
|||||
|
AA4039630
|
Thermo Scientific Chemicals
04039630 |
250 g |
Each for $381.30
|
|
|||||
|
AA40396A1
|
Thermo Scientific Chemicals
040396A1 |
1 kg |
Each for $1,237.48
|
|
|||||
Description
Bismuth(III) chloride oxide is used in pharmaceutical industry, in the production of cosmetics, in chemical and metallurgical industries. It is used as pigment, in manufacture of artificial pearls and as a dry-cell cathodes, in photocatalysts for degradation of organic pollutant, in white-light LEDs, in the manufacture of plastics, in the synthesis of methanol, in semiconductor devices such as CCDs or photodiodes and also used in the production of permanent magnets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7787-59-9 | |
| 261.437 | |
| BWOROQSFKKODDR-UHFFFAOYSA-N | |
| 6328152 | |
| O=[Bi].Cl |
| BiClHO | |
| MFCD00010895 | |
| bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid | |
| oxobismuth;hydrochloride |
Specifications
| (decomposition) | |
| Brown | |
| 50 g | |
| BiOCl | |
| 14,1263 | |
| Insoluble in water,alcohol; Soluble in acids. | |
| O=[Bi].Cl | |
| 261.437 | |
| 260.43 | |
| Odorless | |
| 7.72 g/mL |
| 7787-59-9 | |
| Powder | |
| BiClHO | |
| MFCD00010895 | |
| bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid | |
| BWOROQSFKKODDR-UHFFFAOYSA-N | |
| oxobismuth;hydrochloride | |
| 6328152 | |
| 98% | |
| 2.15 | |
| Bismuth(III) chloride oxide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 232-122-7
RTECSNumber : EB2700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only