missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) Chloride Anhydrous 97.0+%, TCI America™

Catalog No. p-7123011
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
B354625G 25 g
B3546250G 250 g
2 options

Catalog No. B354625G

Supplier: TCI America B354625G

Only null left

Chemical Identifiers

CAS 7787-60-2
Molecular Formula BiCl3
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Specifications

Boiling Point 447°C
Melting Point 230°C
Color Yellow
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 315.33
Percent Purity ≥97.0% (T)
Chemical Name or Material Bismuth(III) Chloride Anhydrous
EINECSNumber (1)-1032
RTECSNumber EB2690000
TSCA Yes