missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) Chloride Anhydrous 97.0+%, TCI America™

Catalog No. B354625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B354625G 25 g
B3546250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B354625G Supplier TCI America Supplier No. B354625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7787-60-2
Molecular Formula BiCl3
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Specifications

Boiling Point 447°C
Melting Point 230°C
Color Yellow
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 315.33
Percent Purity ≥97.0% (T)
Chemical Name or Material Bismuth(III) Chloride Anhydrous
EINECSNumber (1)-1032
RTECSNumber EB2690000
TSCA Yes