missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride, anhydrous, 99.999% (metals basis)

Catalog No. p-7080483
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AA8948506 5 g
AA8948514 25 g
2 options

Catalog No. AA8948506

Supplier: Thermo Scientific Chemicals 08948506

Only null left

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol

It is used as a catalyst in organic chemistry. It is a good source of bismuth and is used to make other bismuth compounds. Also used as a catalyst and to make other bismuth salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-60-2
Molecular Formula BiCl3
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Specifications

Boiling Point ∼430°C (sublimation)
Melting Point 230°C to 232°C
Color White
Physical Form Crystalline
Quantity 5 g
UN Number UN3260
Merck Index 14,1262
Solubility Information Soluble in water.
Formula Weight 315.34
Percent Purity 99.999%
Sensitivity Hygroscopic
Density 4.75 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth (III) chloride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-123-2
RTECSNumber EB2690000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only