missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride, anhydrous, 99.9% (metals basis)

Catalog No. p-4824629
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1711514 25 g
AA1711522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1711514 Supplier Thermo Scientific Chemicals Supplier No. 01711514

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol

Catalysts for organic reactions, preparation of Bi salts.Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-60-2
MDL Number MFCD00003461

Specifications

Boiling Point ca 430°C (sublimation)
Melting Point 230°C to 232°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Molecular Formula BiCl3
Solubility Information Soluble in HCl,HNO3,acetone,ethyl acetate. Insoluble in alcohol. Decomposed by water to BiOCl.Soluble in hydrochloric acid,nitric acid,diethyl ether,ethyl acetate and acetone. Insoluble in alcohol.
Formula Weight 315.34
Odor Irritating
Sensitivity Hygroscopic
Density 4.75
Chemical Name or Material Bismuth(III) chloride
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 232-123-2
RTECSNumber EB2690000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only