missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) bromide, 99% (metals basis)
Description
It catalyzes the formation of cyclic carbonates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 453°C |
| Melting Point | 218°C |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99% (metals basis) |
| Solubility Information | Decomposes in water. |
| Formula Weight | 448.71 |
| Sensitivity | Hygroscopic |
| Density | 5.72 |
| Chemical Name or Material | Bismuth(III) bromide |
Safety and Handling
| Hazard Category | H314-H335 |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 232-121-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only